Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGTGCTTGGCGATCAGCCCAA[C/T]TGAAGGACTGGCTCTGTACTGACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL10 - ADP ribosylation factor like GTPase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317948.1 | 2264 | Intron | NP_001304877.1 | |||
NM_173664.5 | 2264 | UTR 3 | NP_775935.1 | |||
XM_011534529.2 | 2264 | Intron | XP_011532831.1 | |||
XM_011534530.2 | 2264 | Intron | XP_011532832.1 | |||
XM_011534531.2 | 2264 | UTR 3 | XP_011532833.1 | |||
XM_017009372.1 | 2264 | Intron | XP_016864861.1 |
MIR1271 - microRNA 1271 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |