Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTCTTGCAGCCCCTCAAGCTAG[A/G]ATGAAATCCTCAAGAGGATGGTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609551 MIM: 602513 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMAN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LMAN2 - lectin, mannose binding 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RGS14 - regulator of G-protein signaling 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006480.4 | Intron | NP_006471.2 | ||||
XM_005265794.4 | Intron | XP_005265851.1 | ||||
XM_005265795.4 | Intron | XP_005265852.1 |