Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAACCTGCCCTCTGCCCTGGCA[A/C]CCCAGGGACTCACTGCCAAAGACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf31 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C8orf31 - chromosome 8 open reading frame 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173687.3 | 875 | Missense Mutation | ACC,CCC | T,P 70 | NP_775958.1 | |
XM_017013344.1 | 875 | Missense Mutation | ACC,CCC | T,P 82 | XP_016868833.1 | |
XM_017013345.1 | 875 | Missense Mutation | ACC,CCC | T,P 82 | XP_016868834.1 | |
XM_017013346.1 | 875 | Missense Mutation | ACC,CCC | T,P 65 | XP_016868835.1 |
LOC107984097 - uncharacterized protein FLJ40521 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017014161.1 | 875 | Intron | XP_016869650.1 |