Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAATGATTATATAGCTTTTAAAA[A/G]TTATGGAAAAAATACAAATTCTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAA35 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAA35 - N(alpha)-acetyltransferase 35, NatC auxiliary subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321881.1 | Intron | NP_001308810.1 | ||||
NM_001321882.1 | Intron | NP_001308811.1 | ||||
NM_024635.3 | Intron | NP_078911.3 | ||||
XM_005252127.3 | Intron | XP_005252184.1 | ||||
XM_011518903.2 | Intron | XP_011517205.1 | ||||
XM_011518904.2 | Intron | XP_011517206.1 | ||||
XM_017015012.1 | Intron | XP_016870501.1 |