Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCAAGTATGTTGTCCTGGCCACC[A/C]CCTTCTAGGTACCAATGTGAATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615299 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NXNL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NXNL2 - nucleoredoxin-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161625.1 | Intron | NP_001155097.1 | ||||
NM_145283.2 | Intron | NP_660326.2 | ||||
XM_005251727.3 | Intron | XP_005251784.1 | ||||
XM_011518275.2 | Intron | XP_011516577.1 | ||||
XM_011518276.2 | Intron | XP_011516578.1 |