Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAACATCAGATTTATGGGACACC[A/G]TTATGAATACCAACATATGTATAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTUS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTUS2 - microtubule associated tumor suppressor candidate 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033602.2 | Intron | NP_001028774.2 | ||||
NM_015233.5 | Intron | NP_056048.1 | ||||
XM_011535019.2 | Intron | XP_011533321.1 | ||||
XM_011535020.2 | Intron | XP_011533322.1 | ||||
XM_011535021.2 | Intron | XP_011533323.1 | ||||
XM_011535022.2 | Intron | XP_011533324.1 | ||||
XM_017020500.1 | Intron | XP_016875989.1 | ||||
XM_017020501.1 | Intron | XP_016875990.1 | ||||
XM_017020502.1 | Intron | XP_016875991.1 |