Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAAGATTACATAGCTAGTTAGTG[G/T]GAGTGCCATTCTTAGGCTGATGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189973 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELF1 - E74 like ETS transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145353.1 | 3067 | UTR 3 | NP_001138825.1 | |||
NM_172373.3 | 3067 | UTR 3 | NP_758961.1 | |||
XM_005266276.1 | 3067 | UTR 3 | XP_005266333.1 | |||
XM_005266277.3 | 3067 | UTR 3 | XP_005266334.1 | |||
XM_011534950.1 | 3067 | UTR 3 | XP_011533252.1 | |||
XM_011534951.2 | 3067 | UTR 3 | XP_011533253.1 | |||
XM_017020409.1 | 3067 | UTR 3 | XP_016875898.1 | |||
XM_017020410.1 | 3067 | UTR 3 | XP_016875899.1 | |||
XM_017020411.1 | 3067 | UTR 3 | XP_016875900.1 | |||
XM_017020412.1 | 3067 | UTR 3 | XP_016875901.1 | |||
XM_017020413.1 | 3067 | UTR 3 | XP_016875902.1 |