Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTCTGTGTGTGTGTGTGTGTGT[A/G]TATACACGCACATACATCTATACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604861 | ||||||||||||||||||||
Literature Links: |
LATS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LATS2 - large tumor suppressor kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014572.2 | 5083 | UTR 3 | NP_055387.2 | |||
XM_005266342.1 | 5083 | UTR 3 | XP_005266399.1 | |||
XM_011535042.2 | 5083 | UTR 3 | XP_011533344.1 | |||
XM_017020541.1 | 5083 | UTR 3 | XP_016876030.1 | |||
XM_017020542.1 | 5083 | UTR 3 | XP_016876031.1 |