Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTAATTAGTTAACCTATTATTA[A/G]GCTTATGTTATTAAGTTATTAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0226L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0226L - KIAA0226 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286761.1 | Intron | NP_001273690.1 | ||||
NM_001286762.1 | Intron | NP_001273691.1 | ||||
NM_001286763.1 | Intron | NP_001273692.1 | ||||
NM_001286764.1 | Intron | NP_001273693.1 | ||||
NM_001286765.1 | Intron | NP_001273694.1 | ||||
NM_001286766.1 | Intron | NP_001273695.1 | ||||
NM_025113.3 | Intron | NP_079389.2 |