Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGTAGTTCTTCTCGTCAGCGAA[C/T]TGCTCATCATCCGTCCTTTCTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612175 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAB39L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAB39L - calcium binding protein 39 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079670.2 | 1879 | Silent Mutation | CAA,CAG | Q,Q 317 | NP_001073138.1 | |
NM_001287337.1 | 1879 | Silent Mutation | CAA,CAG | Q,Q 317 | NP_001274266.1 | |
NM_001287338.1 | 1879 | Silent Mutation | CAA,CAG | Q,Q 317 | NP_001274267.1 | |
NM_001287339.1 | 1879 | Silent Mutation | CAA,CAG | Q,Q 317 | NP_001274268.1 | |
NM_030925.3 | 1879 | Silent Mutation | CAA,CAG | Q,Q 317 | NP_112187.2 | |
XM_011535254.2 | 1879 | Silent Mutation | CAA,CAG | Q,Q 317 | XP_011533556.1 | |
XM_011535255.2 | 1879 | Silent Mutation | CAA,CAG | Q,Q 317 | XP_011533557.1 | |
XM_011535256.2 | 1879 | Silent Mutation | CAA,CAG | Q,Q 317 | XP_011533558.1 | |
XM_017020785.1 | 1879 | Silent Mutation | CAA,CAG | Q,Q 317 | XP_016876274.1 | |
XM_017020786.1 | 1879 | Silent Mutation | CAA,CAG | Q,Q 317 | XP_016876275.1 | |
XM_017020787.1 | 1879 | Silent Mutation | CAA,CAG | Q,Q 317 | XP_016876276.1 | |
XM_017020788.1 | 1879 | Silent Mutation | CAA,CAG | Q,Q 317 | XP_016876277.1 |