Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGACCCTGAGCTGACCGTGCTTAT[A/G]GATCTGGACAGAGGGGCTCGTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602466 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPRY2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPRY2 - sprouty RTK signaling antagonist 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318536.1 | 875 | Silent Mutation | TCC,TCT | S,S 110 | NP_001305465.1 | |
NM_001318537.1 | 875 | Silent Mutation | TCC,TCT | S,S 110 | NP_001305466.1 | |
NM_001318538.1 | 875 | Silent Mutation | TCC,TCT | S,S 110 | NP_001305467.1 | |
NM_005842.3 | 875 | Silent Mutation | TCC,TCT | S,S 110 | NP_005833.1 |