Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGGTAGGAAATCAGAGCATAAC[A/C]AAGCTTGGGCATGTCTGCTTTACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612450 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HBS1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HBS1L - HBS1 like translational GTPase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145158.1 | 4710 | UTR 3 | NP_001138630.1 | |||
NM_001145207.1 | 4710 | Intron | NP_001138679.1 | |||
NM_006620.3 | 4710 | UTR 3 | NP_006611.1 | |||
XM_017010192.1 | 4710 | Intron | XP_016865681.1 |