Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCCATTCTGAGGACTTTACATC[G/T]GTGTGCAATGGAAGTCATTGAGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604857 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRP54 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRP54 - signal recognition particle 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146282.1 | Intron | NP_001139754.1 | ||||
NM_003136.3 | Intron | NP_003127.1 | ||||
XM_005268024.2 | Intron | XP_005268081.1 | ||||
XM_011537106.1 | Intron | XP_011535408.1 | ||||
XM_017021615.1 | Intron | XP_016877104.1 |