Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGACTACAGTTCCAAAATACAAG[C/T]ACCGACATAATAATAAAATGCAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL28 - kelch like family member 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308112.1 | 3435 | UTR 3 | NP_001295041.1 | |||
NM_017658.4 | 3435 | UTR 3 | NP_060128.2 | |||
XM_005267770.3 | 3435 | UTR 3 | XP_005267827.1 | |||
XM_006720174.3 | 3435 | Intron | XP_006720237.1 | |||
XM_011536847.2 | 3435 | UTR 3 | XP_011535149.1 | |||
XM_011536849.2 | 3435 | UTR 3 | XP_011535151.1 | |||
XM_017021378.1 | 3435 | Intron | XP_016876867.1 |