Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGCCAGCCCTGGCCCACCTGCA[C/G]CCCACATGGAAGCAGAGGGTCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123875 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf80 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf80 - chromosome 14 open reading frame 80 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134875.1 | 590 | Missense Mutation | CCC,GCC | P,A 155 | NP_001128347.1 | |
NM_001134876.1 | 590 | Missense Mutation | CCC,GCC | P,A 155 | NP_001128348.1 | |
NM_001134877.1 | 590 | Missense Mutation | CCC,GCC | P,A 114 | NP_001128349.1 | |
NM_001198983.1 | 590 | Missense Mutation | CCC,GCC | P,A 116 | NP_001185912.1 |
CRIP1 - cysteine rich protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |