Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAAAAATACAGTTCTTGGCCTCA[A/G]GCAAATGGTTATCACTTGGGCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608431 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
G3BP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
G3BP1 - G3BP stress granule assembly factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005754.2 | Intron | NP_005745.1 | ||||
NM_198395.1 | Intron | NP_938405.1 | ||||
XM_006714749.3 | Intron | XP_006714812.1 | ||||
XM_006714750.3 | Intron | XP_006714813.1 | ||||
XM_017008922.1 | Intron | XP_016864411.1 | ||||
XM_017008923.1 | Intron | XP_016864412.1 |
LOC100652758 - uncharacterized LOC100652758 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |