Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACAAAGAACATGGACAGGGTCCT[C/T]CGGTTGTGTAGTCGCCGCTGGACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606647 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARAP3 - ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022481.5 | 3625 | Silent Mutation | CGA,CGG | R,R 1376 | NP_071926.4 | |
XM_005268497.1 | 3625 | Intron | XP_005268554.1 | |||
XM_005268498.1 | 3625 | Silent Mutation | CGA,CGG | R,R 1356 | XP_005268555.1 | |
XM_005268499.1 | 3625 | Silent Mutation | CGA,CGG | R,R 1298 | XP_005268556.1 | |
XM_005268500.3 | 3625 | Intron | XP_005268557.1 | |||
XM_006714792.1 | 3625 | Silent Mutation | CGA,CGG | R,R 1305 | XP_006714855.1 | |
XM_011537677.2 | 3625 | Silent Mutation | CGA,CGG | R,R 1181 | XP_011535979.1 |
FCHSD1 - FCH and double SH3 domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |