Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAGGCCATATCATACCCTGAAGA[A/G]AGGAGCATTTTCCCATGTTGCCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615741 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0141 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0141 - KIAA0141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142603.2 | Intron | NP_001136075.1 | ||||
NM_014773.4 | Intron | NP_055588.3 | ||||
XM_005268547.3 | Intron | XP_005268604.1 | ||||
XM_005268548.3 | Intron | XP_005268605.1 | ||||
XM_005268552.2 | Intron | XP_005268609.1 | ||||
XM_006714812.3 | Intron | XP_006714875.1 | ||||
XM_011537715.1 | Intron | XP_011536017.1 | ||||
XM_011537716.2 | Intron | XP_011536018.1 |