Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCGAGCAAGTTCACTGCTGCCA[A/G]CAGCTCACCTCCAGGGCCTTCCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615741 MIM: 605622 MIM: 605675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0141 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0141 - KIAA0141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142603.2 | 5695 | Intron | NP_001136075.1 | |||
NM_014773.4 | 5695 | Intron | NP_055588.3 |
PCDH12 - protocadherin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016580.3 | 5695 | UTR 3 | NP_057664.1 |
RNF14 - ring finger protein 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |