Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTCCTCCATCTATGAGCTCCTG[A/G]GCCAGGGCCTCCAAGTCAGCTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601078 MIM: 602766 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT82 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRT82 - keratin 82 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033033.3 | 2547 | UTR 3 | NP_149022.3 |
KRT84 - keratin 84 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |