Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACAGAGCAGATGGCCTGGAGAT[C/T]CATGACAAAATCATGGAGGTAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDZRN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDZRN4 - PDZ domain containing ring finger 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164595.1 | 833 | Silent Mutation | ATC,ATT | I,I 275 | NP_001158067.1 | |
NM_013377.3 | 833 | Intron | NP_037509.3 |