Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACTGCTCTTTTCCCCGACCTGC[A/G]GGCTCCCCTCGGTACCAGTTGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 178642 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFTPA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SFTPA2 - surfactant protein A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098668.3 | Intron | NP_001092138.1 | ||||
NM_001320813.1 | Intron | NP_001307742.1 | ||||
NM_001320814.1 | Intron | NP_001307743.1 | ||||
XM_005270128.3 | Intron | XP_005270185.1 | ||||
XM_005270132.3 | Intron | XP_005270189.1 | ||||
XM_011540124.1 | Intron | XP_011538426.1 | ||||
XM_011540125.1 | Intron | XP_011538427.1 | ||||
XM_017016608.1 | Intron | XP_016872097.1 |