Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTTGTTATTGTGGGGAAGAGAG[A/G]TGCAGAATGGCTTCTCAGCGGAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM213A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM213A - family with sequence similarity 213 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243778.1 | Intron | NP_001230707.1 | ||||
NM_001243779.1 | Intron | NP_001230708.1 | ||||
NM_001243780.1 | Intron | NP_001230709.1 | ||||
NM_001243781.1 | Intron | NP_001230710.1 | ||||
NM_001243782.1 | Intron | NP_001230711.1 | ||||
NM_032333.4 | Intron | NP_115709.3 | ||||
XM_011540265.2 | Intron | XP_011538567.1 | ||||
XM_011540266.2 | Intron | XP_011538568.1 | ||||
XM_017016775.1 | Intron | XP_016872264.1 |