Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTTTCCATTTCGTCAGGCGGCT[A/G]TTCTTGTCTGCGTAGCTTTCAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601956 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFRA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GFRA2 - GDNF family receptor alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165038.1 | 1804 | UTR 3 | NP_001158510.1 | |||
NM_001165039.1 | 1804 | UTR 3 | NP_001158511.1 | |||
NM_001495.4 | 1804 | UTR 3 | NP_001486.4 | |||
XM_006716327.3 | 1804 | UTR 3 | XP_006716390.1 | |||
XM_011544484.2 | 1804 | UTR 3 | XP_011542786.1 |