Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAGCATACACTTTCCATGCACA[C/T]TGAGGAGTGCTTTGTTCTTTGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616916 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMAP2 - small ArfGAP2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198978.1 | Intron | NP_001185907.1 | ||||
NM_001198979.1 | Intron | NP_001185908.1 | ||||
NM_001198980.1 | Intron | NP_001185909.1 | ||||
NM_022733.2 | Intron | NP_073570.1 | ||||
XM_011541960.1 | Intron | XP_011540262.1 | ||||
XM_017002093.1 | Intron | XP_016857582.1 |