Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTTGGAGAGAGGCTAAGCGTTG[C/G]GGGGAACCCTGTCCTCAGGCTGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606649 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIVEP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIVEP3 - human immunodeficiency virus type I enhancer binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127714.2 | 8223 | Missense Mutation | CCC,CGC | P,R 2403 | NP_001121186.1 | |
NM_024503.4 | 8223 | Missense Mutation | CCC,CGC | P,R 2404 | NP_078779.2 | |
XM_011541884.2 | 8223 | Missense Mutation | CCC,CGC | P,R 2404 | XP_011540186.1 | |
XM_011541885.2 | 8223 | Intron | XP_011540187.1 | |||
XM_011541886.2 | 8223 | Intron | XP_011540188.1 | |||
XM_011541887.2 | 8223 | Intron | XP_011540189.1 | |||
XM_017001992.1 | 8223 | Missense Mutation | CCC,CGC | P,R 2404 | XP_016857481.1 | |
XM_017001993.1 | 8223 | Missense Mutation | CCC,CGC | P,R 2404 | XP_016857482.1 | |
XM_017001994.1 | 8223 | Missense Mutation | CCC,CGC | P,R 2403 | XP_016857483.1 |