Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCTGGACAAACAGAATAAACAT[A/C]CAAGCTACATTTCGGGTAGGTAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600650 | ||||||||||||||||||||
Literature Links: |
CPT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPT2 - carnitine palmitoyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000098.2 | 508 | Missense Mutation | ACA,CCA | T,P 109 | NP_000089.1 | |
XM_005270484.1 | 508 | Missense Mutation | ACA,CCA | T,P 109 | XP_005270541.1 |