Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTCTTTTTTTAACCAGATTTTA[A/C]AATAACAAGATGGTTCTGTAGTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616773 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIGA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIGA1 - mitoguardin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270384.1 | Intron | NP_001257313.1 | ||||
NM_198549.3 | Intron | NP_940951.1 | ||||
XM_005270846.4 | Intron | XP_005270903.1 | ||||
XM_005270847.4 | Intron | XP_005270904.1 | ||||
XM_005270848.4 | Intron | XP_005270905.1 | ||||
XM_011541422.2 | Intron | XP_011539724.1 | ||||
XM_017001227.1 | Intron | XP_016856716.1 | ||||
XM_017001228.1 | Intron | XP_016856717.1 | ||||
XM_017001229.1 | Intron | XP_016856718.1 | ||||
XM_017001230.1 | Intron | XP_016856719.1 | ||||
XM_017001231.1 | Intron | XP_016856720.1 | ||||
XM_017001232.1 | Intron | XP_016856721.1 |