Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTTCAAAGAGCACACAGCACAT[A/G]AGGCATAGTAATCATCTGTCTTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606254 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEP15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEP15 - 15 kDa selenoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004261.3 | 1170 | UTR 3 | NP_004252.2 | |||
NM_203341.1 | 1170 | UTR 3 | NP_976086.1 |