Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGGAACTACCACTCCTCTGAGG[G/T]TTTCAAGATGTCTGTGTCTACCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC8D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC8D - leucine rich repeat containing 8 family member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134479.1 | Intron | NP_001127951.1 | ||||
NM_018103.4 | Intron | NP_060573.2 | ||||
XM_011541689.1 | Intron | XP_011539991.1 | ||||
XM_017001599.1 | Intron | XP_016857088.1 | ||||
XM_017001600.1 | Intron | XP_016857089.1 | ||||
XM_017001601.1 | Intron | XP_016857090.1 | ||||
XM_017001602.1 | Intron | XP_016857091.1 |