Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATAACAAGACACATATTAATAA[C/T]TGATATATATCTTTTTATTATGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600742 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGFBR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TGFBR3 - transforming growth factor beta receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195683.1 | 6460 | UTR 3 | NP_001182612.1 | |||
NM_001195684.1 | 6460 | UTR 3 | NP_001182613.1 | |||
NM_003243.4 | 6460 | UTR 3 | NP_003234.2 | |||
XM_006710867.2 | 6460 | Intron | XP_006710930.1 | |||
XM_006710868.2 | 6460 | Intron | XP_006710931.1 |