Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGGGCTGGTGGGCCCAATAATG[A/G]GGCCTCTAATGAGCCCCTAATGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610955 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP7D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP7D1 - MAP7 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286365.1 | Intron | NP_001273294.1 | ||||
NM_001286366.1 | Intron | NP_001273295.1 | ||||
NM_018067.4 | Intron | NP_060537.3 | ||||
XM_005271027.1 | Intron | XP_005271084.1 | ||||
XM_006710759.1 | Intron | XP_006710822.1 | ||||
XM_006710761.1 | Intron | XP_006710824.1 |
TRAPPC3 - trafficking protein particle complex 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |