Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACAATTGTGATTTTCTCAAATG[C/T]AGACAATTCAGAGATATTCACAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC39 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC39 - leucine rich repeat containing 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256385.1 | 1985 | Intron | NP_001243314.1 | |||
NM_001256386.1 | 1985 | Intron | NP_001243315.1 | |||
NM_001256387.1 | 1985 | Intron | NP_001243316.1 | |||
NM_144620.3 | 1985 | Intron | NP_653221.1 |
TRMT13 - tRNA methyltransferase 13 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019083.2 | 1985 | UTR 3 | NP_061956.2 | |||
XM_005270945.3 | 1985 | UTR 3 | XP_005271002.1 | |||
XM_005270946.2 | 1985 | UTR 3 | XP_005271003.1 | |||
XM_017001504.1 | 1985 | UTR 3 | XP_016856993.1 | |||
XM_017001505.1 | 1985 | UTR 3 | XP_016856994.1 |