Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAAGGTGAGGGGAGCAGGACTG[A/G]CAGAGGAATAGGTGCCCCAGAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610050 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMPRSS13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMPRSS13 - transmembrane protease, serine 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077263.2 | 3059 | UTR 3 | NP_001070731.1 | |||
NM_001206789.1 | 3059 | UTR 3 | NP_001193718.1 | |||
NM_001206790.1 | 3059 | Intron | NP_001193719.1 | |||
NM_001244995.1 | 3059 | Intron | NP_001231924.1 |