Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTAGTAAGCATCTTGCTACTAC[C/T]GCAAGCTGGTGGCATCCATGAGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608214 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRAMD1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRAMD1B - GRAM domain containing 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCN3B - sodium voltage-gated channel beta subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040151.1 | 3916 | UTR 3 | NP_001035241.1 | |||
NM_018400.3 | 3916 | UTR 3 | NP_060870.1 | |||
XM_011542897.1 | 3916 | Intron | XP_011541199.1 |