Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGACTCAAATGGGTGTGAAGT[A/G]GGAGGGTGGGAAAGCGTGGGTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600734 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf45 - chromosome 11 open reading frame 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256088.1 | 2022 | UTR 3 | NP_001243017.1 | |||
NM_145013.2 | 2022 | UTR 3 | NP_659450.1 | |||
XM_011542654.2 | 2022 | UTR 3 | XP_011540956.1 | |||
XM_011542655.2 | 2022 | UTR 3 | XP_011540957.1 |
KCNJ5 - potassium voltage-gated channel subfamily J member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000890.3 | 2022 | Intron | NP_000881.3 | |||
XM_011542809.2 | 2022 | Intron | XP_011541111.1 | |||
XM_011542810.2 | 2022 | Intron | XP_011541112.1 |