Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGCCCCAGCTTCAACAACCTCC[A/G]AAGGCGACAATGGAGGAAGACAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSGA10IP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSGA10IP - testis specific 10 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152762.2 | 1139 | Missense Mutation | CAA,CGA | Q,R 303 | NP_689975.2 | |
XM_011544887.2 | 1139 | Missense Mutation | CAA,CGA | Q,R 303 | XP_011543189.1 | |
XM_011544888.2 | 1139 | Missense Mutation | CAA,CGA | Q,R 167 | XP_011543190.1 |