Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAACCTGAATCTGGGCAGAAAGC[C/T]GATGCCAATGACAGAGACAAGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613664 MIM: 603890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMTNL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMTNL1 - smoothelin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105565.2 | 458 | Silent Mutation | GCC,GCT | A,A 152 | NP_001099035.2 | |
XM_005273819.3 | 458 | Silent Mutation | GCC,GCT | A,A 152 | XP_005273876.1 |
UBE2L6 - ubiquitin conjugating enzyme E2 L6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |