Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCCACCCCCAACACCAGCCTGC[C/T]TGTCACATGTTGTGACACTCTTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608403 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MS4A10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MS4A10 - membrane spanning 4-domains A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206893.3 | Intron | NP_996776.2 | ||||
XM_011544989.1 | Intron | XP_011543291.1 | ||||
XM_011544991.2 | Intron | XP_011543293.1 |
MS4A15 - membrane spanning 4-domains A15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |