Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGAGCAACAGGGCTCAGCACAG[A/G]TTTGCAGAGCATCAACATAACTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604433 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNE3 - potassium voltage-gated channel subfamily E regulatory subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005472.4 | 2901 | UTR 3 | NP_005463.1 | |||
XM_011544713.2 | 2901 | Intron | XP_011543015.1 | |||
XM_017017047.1 | 2901 | Intron | XP_016872536.1 | |||
XM_017017048.1 | 2901 | Intron | XP_016872537.1 | |||
XM_017017049.1 | 2901 | Intron | XP_016872538.1 | |||
XM_017017050.1 | 2901 | Intron | XP_016872539.1 | |||
XM_017017051.1 | 2901 | Intron | XP_016872540.1 | |||
XM_017017052.1 | 2901 | Intron | XP_016872541.1 |