Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTTCCTCTCCCAGCAGCTGTAA[A/G]CGCCCTACAGTTTGCATGGGAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600835 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf142 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C10orf142 - chromosome 10 open reading frame 142 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258000.1 | 194 | Silent Mutation | AAA,AAG | K,K 61 | NP_001244929.1 |
CXCL12 - C-X-C motif chemokine ligand 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000609.6 | 194 | Intron | NP_000600.1 | |||
NM_001033886.2 | 194 | Intron | NP_001029058.1 | |||
NM_001178134.1 | 194 | Intron | NP_001171605.1 | |||
NM_001277990.1 | 194 | Intron | NP_001264919.1 | |||
NM_199168.3 | 194 | Intron | NP_954637.1 |