Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTATAAATTCATTCATTCAACAG[C/G]TATTTACTGAGTGTCTGTTCCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610559 MIM: 194529 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C10orf25 - chromosome 10 open reading frame 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039380.3 | 1137 | UTR 3 | NP_001034469.2 | |||
XM_017015895.1 | 1137 | Intron | XP_016871384.1 |
RASSF4 - Ras association domain family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF22 - zinc finger protein 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |