Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAAGATAGCTTAGAATTAAGAG[C/T]TAACTTCTAGTCAGTATTGGTAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927100 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927100 - uncharacterized LOC101927100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMED7 - transmembrane p24 trafficking protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181836.5 | 2505 | UTR 3 | NP_861974.1 |
TMED7-TICAM2 - TMED7-TICAM2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164468.3 | 2505 | Intron | NP_001157940.1 | |||
NM_001164469.3 | 2505 | Intron | NP_001157941.1 |