Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAAAAACATTTGTTAGAATCCTA[C/T]AGATATTAAGTTGAACCATTTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604103 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYOT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYOT - myotilin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135940.1 | Intron | NP_001129412.1 | ||||
NM_001300911.1 | Intron | NP_001287840.1 | ||||
NM_006790.2 | Intron | NP_006781.1 | ||||
XM_017010060.1 | Intron | XP_016865549.1 | ||||
XM_017010061.1 | Intron | XP_016865550.1 | ||||
XM_017010062.1 | Intron | XP_016865551.1 |