Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTCTTTTTAAATTAGAACTCTC[C/T]TCATTTGTCTTTCCCTTTTTATAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604383 MIM: 605284 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFI1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GFI1B - growth factor independent 1B transcriptional repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135031.1 | Intron | NP_001128503.1 | ||||
NM_004188.6 | Intron | NP_004179.3 | ||||
XM_006717297.3 | Intron | XP_006717360.1 | ||||
XM_011519068.2 | Intron | XP_011517370.1 | ||||
XM_011519069.2 | Intron | XP_011517371.1 | ||||
XM_011519070.2 | Intron | XP_011517372.1 | ||||
XM_017015175.1 | Intron | XP_016870664.1 | ||||
XM_017015176.1 | Intron | XP_016870665.1 |
MIR548AW - microRNA 548aw | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSC1 - tuberous sclerosis 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |