Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGTGGCCACGTGAGCCTCTGAG[G/T]ATTCAGTGTCCCCTGCAGGTCCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604349 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIF1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIF1L - allograft inflammatory factor 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185095.1 | Intron | NP_001172024.1 | ||||
NM_001185096.1 | Intron | NP_001172025.1 | ||||
NM_031426.3 | Intron | NP_113614.1 | ||||
XM_017015177.1 | Intron | XP_016870666.1 |
LAMC3 - laminin subunit gamma 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |