Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACGGGCGGCAGTGTCGTCAGTT[A/G]CAGGGGTGCAGTCCAAGGCCCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607904 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNA1H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNA1H - calcium voltage-gated channel subunit alpha1 H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005407.1 | Intron | NP_001005407.1 | ||||
NM_021098.2 | Intron | NP_066921.2 | ||||
XM_005255652.4 | Intron | XP_005255709.1 | ||||
XM_006720963.3 | Intron | XP_006721026.1 | ||||
XM_006720964.3 | Intron | XP_006721027.1 | ||||
XM_006720965.3 | Intron | XP_006721028.1 | ||||
XM_006720967.3 | Intron | XP_006721030.1 | ||||
XM_006720968.3 | Intron | XP_006721031.1 | ||||
XM_011522724.2 | Intron | XP_011521026.1 | ||||
XM_011522727.2 | Intron | XP_011521029.1 | ||||
XM_017023819.1 | Intron | XP_016879308.1 | ||||
XM_017023820.1 | Intron | XP_016879309.1 | ||||
XM_017023821.1 | Intron | XP_016879310.1 |