Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGCAAATACATTAACATTGGGG[G/T]TTTTTTAATACTTCTGTATCTTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602053 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLF6 - Kruppel like factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160124.1 | 2612 | UTR 3 | NP_001153596.1 | |||
NM_001160125.1 | 2612 | UTR 3 | NP_001153597.1 | |||
NM_001300.5 | 2612 | UTR 3 | NP_001291.3 |