Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCACCATGCTCAGACATAGAGGT[A/C]ATGGTTCTTAATCTGGTGTGTGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300935 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRRG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRRG1 - proline rich and Gla domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000950.2 | Intron | NP_000941.1 | ||||
NM_001142395.1 | Intron | NP_001135867.1 | ||||
NM_001173486.1 | Intron | NP_001166957.1 | ||||
NM_001173489.1 | Intron | NP_001166960.1 | ||||
NM_001173490.1 | Intron | NP_001166961.1 |