Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGTGACGAGAGCTTTTTGCCTTA[C/T]ATATTGATTATATATTATAAATATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300185 MIM: 300456 | ||||||||||||||||||||
Literature Links: |
AKAP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKAP4 - A-kinase anchoring protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCNB3 - cyclin B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033031.2 | Intron | NP_149020.2 | ||||
NM_033670.2 | Intron | NP_391990.1 | ||||
XM_017029913.1 | Intron | XP_016885402.1 | ||||
XM_017029914.1 | Intron | XP_016885403.1 | ||||
XM_017029915.1 | Intron | XP_016885404.1 | ||||
XM_017029916.1 | Intron | XP_016885405.1 | ||||
XM_017029917.1 | Intron | XP_016885406.1 | ||||
XM_017029918.1 | Intron | XP_016885407.1 |